Mouse Recombinant CNTF (from E. coli)
Fournisseur: Shenandoah Biotechnology
SHBT200-90AF-100UGEA 0 EUR InStock
SHBT200-90AF-100UG SHBT200-90AF-500UG SHBT200-90AF-1MG SHBT200-90AF-5UG SHBT200-90AF-20UG
Mouse Recombinant CNTF (from E. coli)
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis.
  • Animal-free
  • Plays an important protective role during nervous system injury
  • Contains 198 amino acids
  • Produced from genetically modified E. coli
Information de livraison: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Attention: This product is for research use only and is not for use in humans.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité