H32091.14
H32091.22
H32091.14EA
40.8
EUR
InStock
H32091.14
H32091.22
N,N-Dibenzylaniline ≥99%
N,N-Dibenzylaniline
Purity: 99%
CAS No.: 91-73-6
Molecular Formula: C20H19N
Formula Weight: 273.37
Melting Point: 69-71°
Boiling Point: 180°/1mm
MDL No.: MFCD00022015