X-ray and metal detectable spoons
Fournisseur: Sampling Systems
SAMPM1006B-10EA 8.6 EUR InStock
SAMPM1006B-10
X-ray and metal detectable spoons
Cuillères
The X-ray and metal detectable spoons are ideal for use in food and pharmaceutical production and sampling. The spoons can be used with both powders and liquids. They are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder or liquid in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Available in 10 ml volume
  • Full batch traceability
To ensure the cleanliness of the spoons they are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
These PP spoons conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Conditionnement: Supplied individually bagged.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité