Guardian 5000 magnetic hotplate stirrers
Fournisseur: OHAUS
30500523 30500553 30500563
OHAU30500523 OHAU30500553 OHAU30500563
Guardian 5000 magnetic hotplate stirrers
Agitateurs Agitateurs magnétiques chauffants
Reliable magnetic hotplate stirrer with exceptional safety features for everyday use.
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Easy to read LCD display
  • Designed for durability and reliability with SmartHousing™
Guardian 5000™ magnetic hotplates stirrers are designed for safety and simplicity in all research, academic and industrial applications.
The design ensures proper mixing and temperature control of the top plate or your sample when using the optional temperature probe.
Heating models include a Single Point Calibration feature for multiple setpoints. Choice of models with differing plate sizes to suit different everyday heating and stirring applications.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité