Alco liquid 80
Fournisseur: Local
1880345-12X1L 1880345-5L
LOCA1880345-12X1LEA 158 EUR InStock
LOCA1880345-12X1L LOCA1880345-5L
Alco liquid 80
Produits nettoyants Désinfectants
Alco Liquid 80 is a disinfectant product with an 80% alcohol concentration (fully denatured ethanol, in accordance with Regulation (EC) No. 3199/93), which can be used to combat a wide range of micro-organisms. the high concentration of active ingredients destroys the cell membrane of the microorganisms.
Alco Liquid 80 is registered in accordance with the Royal Decree of April 4, 2019 regarding the making available on the market and the use of biocides (BE-REG-00556).
Alco Liquid 80 can be used as a hand disinfectant (PT 1) and cleaning agent (PT 2), and can be used in the context of the control of Covid-19.
Information de livraison: Each pack of 12×1 L bottles is supplied with two sprayheads.
Attention: Use biocides safely. Before use, read the label and product information.
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité