APOSBIM109-50MGEA
531
EUR
InStock
APOSBIM109-50MG
Sulphosuccinimidyl-4-[2-(4,4-dimethoxytrityl)]butyrate
Sulphosuccinimidyl-4-[2-(4,4-dimethoxytrityl)]butyrate
Detects immobilized amines
Ref: Gaur.R.K. and Gupta,K.C. (1989) Anal.Biochem. 180, 253-258