Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
OR70067-1G OR70067-5G OR70067-10G
APOSOR70067-1GEA 21.9 EUR InStock
APOSOR70067-1G APOSOR70067-5G APOSOR70067-10G
2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected
2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected
Formula: C₇H₁₃NO₅S
MW: 223.247 g/mol
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD20527196
CAS Number: 459817-82-4
UN: 3077
ADR: 9+EHS III
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty