Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nonafluoro-tert-butyl isobutyrate

Supplier: Apollo Scientific
Warning

PC2892-1G
APOSPC2892-1GEA 411 EUR InStock
APOSPC2892-1G
Nonafluoro-tert-butyl isobutyrate
Nonafluoro-tert-butyl isobutyrate


Formula: C₈H₇F₉O₂
MW: 306.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08741387
CAS Number: 914637-43-7
UN: 1993
ADR: 3 III
Order Now

 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 016 385 011
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 016 385 011.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Customer Service at vwrbe@be.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Products marked with this symbol have been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 016/385 011.
Collapse All
Pack type Pk VWR Catalog Number Quantity
Bottle 1 g APOSPC2892-1G

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory, Enter 'x' if unknown for VWR Chemicals

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty

Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.

Pour en savoir plus, consultez notre Politique de Confidentialité..