Synonyms:
1H,1H,3H-Tétrafluoro-1-propanol
APOSPC6864-2.5KGEA
379
EUR
InStock
APOSPC6864-2.5KG
APOSPC6864-500G
APOSPC6864-100G
2,2,3,3-Tétrafluoro-1-propanol 98%
2,2,3,3-Tétrafluoro-1-propanol
- Surface tension 27.6 dynes/cm 200C
- Dissociation constant Ka=4.6 x 10-12