85716.180EA
96.9
EUR
InStock
85716.180
Eau de brome Reag. Ph. Eur. 1012402
Eau de brome
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.