Laboratory balances verified, France country-specific, Secura®
Fournisseur: Sartorius Balances
SECURA11031CFR SECURA324-1CFR
SARBSECURA11031CFREA 6580 EUR InStock
SARBSECURA11031CFR SARBSECURA324-1CFR
Laboratory balances verified, France country-specific, Secura®
Balances
Secura® gives the security and peace of mind of knowing that you have done everything right. Besides providing highly accurate weighing results and operating convenience, Secura® also features built-in protection systems for complete reliability and regulatory compliance – the safe and secure way.
  • Reduce the risk with integrated and intelligent protection systems
  • Real-time guidance prompts for levelling and 100% traceable, clear documentation (Cal Audit Trail) with sample and batch identifiers make laboratory work more efficient
  • Integrated protection systems for minimizing the risk of obtaining incorrect results
  • Real-time alarm messaging and interactive user guidance for levelling to prevent inaccurate results due to an unlevelled balance
Secura® features SQmin for automatic detection and active monitoring to ensure you are within the acceptable USP operating range for reliable results. Fully automatic internal adjustment function-isoCAL, gives highest possible accuracy at all times. Models have standard password security features for protection against tampering and unauthorized access and provide safe and clear documentation by sample and batch identifiers for keeping track of weighing data as well as GLP/GMP-compliant printouts.
Cal Audit Trail for easy, traceable documentation of all calibration and adjustment data. Integrated PC direct function and Plug and Work connectivity to a Sartorius printer.
Information de livraison: Supplied with universal power adapter, in-use cover, application guide, dust cover for analytical balances and a draft shield ring for 0,1 mg balances.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité