Big Blue Filter Cartridge Housings, Polypropylene
Fournisseur: Big Blue
MFLX01508-75EA 80.1 EUR InStock
MFLX01508-75 MFLX01508-65 MFLX01507-76 MFLX01508-67 MFLX01507-77 MFLX29802-01 MFLX01508-71 MFLX29802-10 MFLX01508-70 MFLX29802-00 MFLX29802-11 MFLX01507-78
Big Blue Filter Cartridge Housings, Polypropylene
Filtres Cartouches filtrantes
Flow rates up to 50 GPM in a compact, relatively low-cost package.
  • Constructed of durable Polypropylene
These systems offer up to four times the filtration area than standard cartridge systems, which makes them ideal for high-flow processes. A wide range of cartridges is offered for numerous applications. Order housings and cartridges separately.
Housing and cap are constructed of durable polypropylene and sealed with a Buna-N O-ring. Housing is suitable to 100 °F (37 °C); maximum operating pressure is 100 psi (6.9 bar).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité