Silastic® Laboratory Transfer Pump Tubing
Fournisseur: Avantor Fluid Handling
MFLX96115-08EA 112 EUR InStock
MFLX96115-08 MFLX96115-18 MFLX96115-28 MFLX96115-06 MFLX96115-30 MFLX96115-40 MFLX96115-38 MFLX96115-16 MFLX96115-26 MFLX96115-04 MFLX96115-14 MFLX96115-24 MFLX96115-02 MFLX96115-12 MFLX96115-34 MFLX96115-44 MFLX96115-00 MFLX96115-22 MFLX96115-32 MFLX96115-10 MFLX96115-20
Silastic® Laboratory Transfer Pump Tubing
Tuyaux Transfer Tubing
Platinum-cured tubing for a wide range of applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotech, filtration, and environmental analysis
Characteristics: Platinum-cured, nonpyrogenic, bacteria and fungus-resistant, extremely low extractables, and does not impart a taste or odor
Certifications: Exceeds USP Class VI Plastics Test requirements, elastomer and tubing are manufactured under Bulk Pharmaceutical cGMPs in a FDA compliant, ISO-9002-certified facility
Temperature range: –65 °F to 480 °F (–54 °C to 249 °C)
Sterilize: Autoclave or radiation.
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité