Female Threaded Cap Fittings, PTFE
Fournisseur: Avantor Fluid Handling
MFLX34030-61EA 73.7 EUR InStock
MFLX34030-61 MFLX34030-62 MFLX34030-65 MFLX34030-64 MFLX34030-66
Female Threaded Cap Fittings, PTFE
Raccords et connecteurs
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité