(-)-Epigallocatechin gallate ≥95% (par HPLC), Calbiochem®, Sigma-Aldrich®
Fournisseur: Merck Millipore Calbiochem
Attention
M-Clarity™ (Niveau de qualité) = MQ100
324880-10EA 121 EUR InStock
324880-10
(-)-Epigallocatechin gallate ≥95% (par HPLC), Calbiochem®, Sigma-Aldrich®
(-)-Epigallocatechin gallate
One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties
Arrests cell cycling at G0/G1 phases and induces apoptosis in a dose-dependent manner. EGCG has been shown to inhibit PMA-induced skin thickening, and the activation of protein kinase C. Also inhibits the activation of ornithine decarboxylase, and activation of interleukin-1[alpha] mRNA and protein expression. Inhibits inducible nitric oxide synthase (iNOS) gene expression and enzyme activity and inhibits peroxynitrite-mediated formation of 8-oxodeoxyguanosine and 3-nitrotyrosine. Also strongly and directly inhibits telomerase.
Conditionnement: Packaged under inert gas
Formule: C₂₂H₁₈O₁₁
MW: 458,38 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00075940
Numéro CAS: 989-51-5
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité