AMAS (N-Succinimidyl maleimidoacetate), Pierce™
Fournisseur: Thermo Fisher Scientific
Synonyms: AMAS
Pierce™
PIER22295EA 173 EUR InStock
PIER22295
AMAS (N-Succinimidyl maleimidoacetate), Pierce™
AMAS (N-Succinimidyl maleimidoacetate)
Thermo Scientific Pierce AMAS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a very short spacer arm (4.4 angstroms).
  • Reactive groups: NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Shortest of aliphatic spacer series which includes AMAS, BMPS, GMBS and EMCS
  • NHS ester end couples with primary amines at pH 7-9, forming stable amide bonds
  • Maleimide reacts with -SH groups at pH 6.5-7.5, forming stable thioether linkages
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO)
  • Very short aliphatic spacer has low potential for eliciting an immune response
Attention: For Research Use Only. Not for use in diagnostic procedures.
Formule: C₁₀H₈N₂O₆
MW: 252,18 g/mol
Numéro MDL: MFCD00133523
Numéro CAS: 55750-61-3
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité