Adapter Fittings, Compression to Male Threaded, Elbow, Jaco Manufacturing®
Fournisseur: Jaco Manufacturing
MFLX30805-45BLKEA 258 EUR InStock
MFLX30805-45BLK MFLX30805-42BLK MFLX30805-40BLK
Adapter Fittings, Compression to Male Threaded, Elbow, Jaco Manufacturing®
Raccords et connecteurs
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression and threaded fittings offer a more permanent installation. Choose the size and material that suit your application needs.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité