Adapter Fittings, Compression to Male Threaded, Straight, Jaco Manufacturing®
Fournisseur: Jaco Manufacturing
MFLX06385-10BLKEA 115 EUR InStock
MFLX06385-10BLK MFLX06385-05BLK MFLX30805-27BLK MFLX06385-50BLK MFLX30805-32BLK MFLX06385-70BLK MFLX30805-36BLK MFLX06385-20BLK MFLX30805-33BLK MFLX30805-38BLK MFLX30805-29BLK MFLX06382-38BLK MFLX06385-40BLK MFLX06382-36BLK MFLX06385-60BLK
Adapter Fittings, Compression to Male Threaded, Straight, Jaco Manufacturing®
Raccords et connecteurs
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité