Moticam 4000 digital camera, HDMI and USB outputs
Fournisseur: Motic
Moticam
MOTI1100600101522EA 1600 EUR InStock
MOTI1100600101522
Moticam 4000 digital camera, HDMI and USB outputs
Caméras
The Moticam 4000 is designed for efficiency and to output fast live images at 4K (3840×2160 px). Connect it through the HDMI port to a screen and display your live image. With the use of the on-board software and a USB mouse, you can make live measurements, notes, image adjustments and more, directly on the HDMI screen, without the need for a computer.
  • True 4K imaging - crisp and clear images displayed in your screen via HDMI
  • On-board software, controlled with a USB mouse connected to the camera
  • Live calibration and measurements, image adjustments, annotations, and more
  • Multi-output: display your samples simultaneously in different devices
  • All-in-one box - all the accessories and software you might need to digitise your samples
Meet our newest HDMI standalone camera: The Moticam 4000. Designed primarily to connect to a 4K monitor via HDMI, it also features USB ports, allowing you to display your live image on your computer. Both HDMI screen and computer can be used at the same time to visualise your samples.
Discover our newest Moticam 4000, designed for efficiency and to output fast live images at 4K (3840×2160 px). Connect it through the HDMI port to a screen and display your live image. With the use of the on-board software and a USB mouse, you can make live measurements, notes, image adjustments and more, directly on the HDMI screen, without the need for a computer. Thanks to its integrated SD card slot and USB ports, saving images won't require a computer, making it easy to save and organise your images and videos. You can also connect your camera through USB to a Windows, or Mac computer, and even display your samples simultaneously.
Following our All-In-One-Box style, with the included accessories you can convert almost any conventional microscope into a digital microscope. This Moticam has the necessary equipment, including the software, to help you to turn your images into knowledge.
Let the Moticam 4000 built-in software guide you in viewing live images, making measurements, image adjustments, or editing them. You can also capture images or record videos directly from a screen with your mouse, storing them in an SD card or a USB stick. If you wish to work with our well-known software Motic Images Plus 3,1, which comes included in the package, connect the camera through USB to your computer.
Thanks to its integrated SD card slot and USB ports, saving images won't require a computer. You can also connect your camera through USB to a Windows, or Mac computer, and even display your samples simultaneously.
Informations de commande: C-mount camera; the C-mount adapter must be purchased separately to mount on a trinocular microscope.
Information de livraison: CS ring adaptor, focusable lens, 30 and 38 mm eyepiece adapters, Motic 4-dot calibration slide, macro calibration card, macro tube, universal power supply, HDMI cable, USB cable, USB mouse and USB pendrive with Motic Images Plus 3,1.
Conditionnement: Delivered in one box of 310×260×270 mm = 1,6 kg.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité