Value Plastics® Fittings, Y-Adapter, Hose Barb Union, PVDF
Fournisseur: Avantor Fluid Handling
Value Plastics®
MFLX40617-21BLKEA 162 EUR InStock
MFLX40617-21BLK MFLX40617-71BLK MFLX40617-70BLK
Value Plastics® Fittings, Y-Adapter, Hose Barb Union, PVDF
Raccords et connecteurs
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock - most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, or food processing.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité