Value Plastics® Fitting, Reducing Barb ¹/₄" ID to ¹/₂"
Fournisseur: Avantor Fluid Handling
Value Plastics®
MFLX40617-14BLKEA 319 EUR InStock
MFLX40617-14BLK
Value Plastics® Fitting, Reducing Barb ¹/₄" ID to ¹/₂"
Raccords et connecteurs
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperature range −80 to 265 °F (−62 to 129 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip the flexible tubing or hose onto the barb to create a secure seal.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité