HDAC6 Inhibitor Screening Kit (Fluorometric)
Fournisseur: BioVision
ABCAAB283385-100EA 650 EUR InStock
ABCAAB283385-100
HDAC6 Inhibitor Screening Kit (Fluorometric)
Tests Tests enzymatiques
High-throughput compatible, quick & easy screening for HDAC6 ligands; Includes hu recombinant HDAC6.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of HDAC6 inhibitors
  • Kit includes Tubacin, an HDAC6 inhibitor, and a stable, recombinant human HDAC6
Histone Deacetylase 6 (HDAC6, EC 3.5.1.98), is a class IIb HDAC enzyme that deacetylates an ε-N-acetyl lysine of histone and non-histone protein substrates. It is a unique member of HDAC family as it contains two deacetylase domains that are proposed to function independently of each other. HDAC6 can shuttle between the nucleus and cytoplasm, suggesting potential extra-nuclear functions by regulating the acetylation status of non-histone substrates. HDAC6 also affects transcription and translation by regulating the heat-shock protein 90 (Hsp90) and stress granules.
Elevated HDAC6 activity is associated with cell motility and catalyzes increases α-tubulin deacetylation, thus influencing cancer cell metastasis. In addition, mutations in HDAC6 gene have been associated with Alzheimer's disease. HDAC6-selective inhibitors are considered as promising targets against autoimmune, inflammatory and oncology diseases such as breast and ovarian cancer. BioVision’s HDAC6 Inhibitor Screening Kit utilizes deacetylase activity of HDAC6 towards a synthetic acetylated-peptide substrate resulting in the release of an AFC fluorophore, which can be easily quantified using a conventional microplate reader.
In the presence of an HDAC6-selective inhibitor (Tubacin), the deacetylase activity of HDAC6 is reduced/abolished resulting in decrease or total loss of the fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of HDAC6.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité