Macrophage Generation Medium DXF
Fournisseur: MARKET SOURCE PART PROCESS
C-28055 C-28056
MSPPC-28055EA 0 EUR InStock
MSPPC-28055 MSPPC-28056
Macrophage Generation Medium DXF
Milieux de culture cellulaire
The PromoCell Macrophage Generation Medium was developed for easy and efficient generation of M1- or M2-polarised macrophages from freshly isolated peripheral blood monocytes.
The PromoCell M1-Macrophage Generation Medium DXF allows for the generation of M1-polarised macrophages, whereas the M2-Macrophage Generation Medium DXF produces M2-polarised macrophages. The chemically defined and Xeno-free media formulations are free of all animal derived components and substances of human origin with Human Serum Albumin as the only exception. The Macrophage Base Medium does not include cytokines and must therefore be supplemented according to the user’s individual needs. The Monocyte Attachment Medium allows for the efficient adherence of fresh monocytes as well as for adherence selection of monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes prior to macrophage differentiation is not necessary.
Recommended for Human Mononuclear Cells, freshly isolated, Human Monocytes, freshly isolated.
A detailed step-by-step protocol is available also, which allows optional user-customised macrophage activation. In combination with the Monocyte Attachment Medium, freshly isolated peripheral blood mononuclear cells can also be directly used as a starting material.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité