Monocyte Attachment Medium
Fournisseur: PromoCell
MSPPC-28051EA 0 EUR InStock
MSPPC-28051
Monocyte Attachment Medium
Milieux de séparation cellulaire
The Monocyte Attachment Medium allows for the efficient adherence selection of Monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health.
Recommended for Human Mononuclear Cells, freshly isolated.
Thus, the time-consuming and costly immunomagnetic purification of monocytes can be skipped. With properly performed washing steps (see Instruction Manual) large numbers of 80 to 90% pure monocytes can be obtained with the PromoCell Monocyte Attachment Medium. Typical applications are generation of dendritic cells/macrophages as well as phagocytosis assays.
Informations de commande: The Medium is ready-to-use and does not need further supplementation.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité