Macrophage Detachment Solution DXF
Fournisseur: PromoCell
MSPPC-41330EA 0 EUR InStock
MSPPC-41330
Macrophage Detachment Solution DXF
Milieux de séparation cellulaire
The Macrophage Detachment Solution DXF is a non-enzymatic reagent especially optimised for the gentle detachment of strongly adherent macrophages.
  • Animal component-free
Macrophages are strongly adherent and are not effectively dislodged from culture surfaces by standard enzymatic cell dissociation reagents. Indeed, these cells are highly sensitive to improper dissociation procedures. It was especially designed for the gentle release of adherent macrophages and guarantees the best possible cell viability, even after prolonged exposure times. Unlike enzyme-based solutions the PromoCell Macrophage Detachment Solution DXF does not alter cell surface proteins and neutralisation is not necessary.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité