Cleanroom Nitrile Gloves, Extended Cuff, Sterile, CRN+
Fournisseur: Isofield
112-6490EA 508 EUR InStock
112-6490 112-6491 112-6492 112-6493
Cleanroom Nitrile Gloves, Extended Cuff, Sterile, CRN+
Gants
The Isofield CRN+ sterile nitrile extended cuff glove offers excellent elbow length protection for GMP Grade A, ISO Class 4 cleanrooms and above.
  • ISO Class 4 compatible
  • Sterile cleanroom nitrile
  • Extra length (400 mm)
  • Low endotoxin
  • Tested against Chemotherapy
  • Ambidextrous
  • Anti-static
Triple washed in de-ionized water, the low endotoxin CRN+ glove has been tested for chemotherapy against ASTMD 6978-05. Anti-static and double-donnable.
Certifications: PPE Cat III, Type B certified.
Conditionnement: Each pair is double packaged in a wallet within a pouch, 10 pouches per bag, 20 bags per case (200 pairs).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité