Safety-Walk™ 3270E Cushion Mattings
Fournisseur: 3M
MMMA3270E930EA 935 EUR InStock
MMMA3270E930
Safety-Walk™ 3270E Cushion Mattings
Tapis de sol Tapis anti-fatigue
Safety Walk™ Light duty cushion matting 3270E is made from continuous coiled vinyl filaments that provide comfort and support across the bottom of the shoe, contributing to a reduction in leg and back fatigue. The textured top helps improve traction and allows debris and liquid to stay on top of the mat for easy cleanup.
  • Anti-fatigue cushion matting which helps reduce leg and back fatigue
  • Textured top helps improving traction for safer footing
  • Supplied with edging to allow wheeled traffic
  • Withstands most industrial and cleaning chemicals
  • Special vinyl coils to withstand most industrial and cleaning chemicals
The mat features integral ramped edging on all sides to enable wheeled traffic movement and pedestrian safety. The product is suitable for dry work areas such as assembly lines, packing areas, and security posts.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité