ISAspher HPLC Columns, C18 BDS
Fournisseur: Isera
ISAspher
ISERA112-C23E30EA 94 EUR InStock
ISERA112-C23E30 ISERA113-C29D00 ISERA111-C23H00 ISERA111-C25K00 ISERA111-C29Q00 ISERA112-C23G00 ISERA111-C29M00 ISERA111-C23L00 ISERA113-C25K00 ISERA111-C25C00 ISERA112-C25F00 ISERA111-C25G00 ISERA111-C23D00 ISERA111-C29E00 ISERA112-C25B00 ISERA112-C23C00 ISERA111-C23X00 ISERA112-C25C30 ISERA112-C29E30 ISERA112-C29A30 ISERA111-C23P00 ISERA111-C25S00 ISERA111-C29Y00 ISERA111-C23T00 ISERA111-C25W00 ISERA111-C29U00 ISERA112-C25G30 ISERA112-C29C00 ISERA111-C29C00 ISERA113-C29A00 ISERA111-C25J00 ISERA111-C29P00 ISERA111-C23G00 ISERA111-C29L00 ISERA111-C25N00 ISERA113-C25L00 ISERA111-C23K00 ISERA112-C25E00 ISERA111-C29H00 ISERA112-C23B00 ISERA111-C25B00 ISERA113-C29J00 ISERA113-C25H00 ISERA112-C29G00 ISERA111-C29D00 ISERA112-C25A00 ISERA112-C23F00 ISERA111-C25F00 ISERA113-C25D00 ISERA111-C23C00 ISERA111-C25Z00 ISERA112-C25B30 ISERA111-C23W00 ISERA111-C25R00 ISERA111-C29X00 ISERA111-C25V00 ISERA111-C29T00 ISERA111-C23S00 ISERA112-C23C30 ISERA112-C25F30 ISERA112-C23G30 ISERA111-C29B00 ISERA112-C29B00 ISERA113-C29B00 ISERA111-C23F00 ISERA111-C29S00 ISERA111-C25M00 ISERA111-C23J00 ISERA112-C23A00 ISERA113-C29K00 ISERA111-C25A00 ISERA111-C29K00 ISERA112-C23E00 ISERA111-C23B00 ISERA111-C25E00 ISERA111-C29G00 ISERA112-C29F00 ISERA112-C29G30 ISERA112-C25A30 ISERA111-C25Y00 ISERA111-C23V00 ISERA112-C29C30 ISERA111-C23Z00 ISERA111-C25Q00 ISERA111-C23N00 ISERA111-C23R00 ISERA111-C29W00 ISERA111-C25U00 ISERA112-C23B30 ISERA112-C25E30 ISERA112-C23F30 ISERA111-C29A00 ISERA113-C25A00 ISERA112-C29A00 ISERA111-C29R00 ISERA111-C25H00 ISERA113-C25J00 ISERA111-C23M00 ISERA111-C29N00 ISERA111-C25L00 ISERA111-C23A00 ISERA113-C29L00 ISERA112-C25G00 ISERA111-C29J00 ISERA113-C29H00 ISERA111-C23E00 ISERA113-C25B00 ISERA112-C29E00 ISERA111-C29F00 ISERA112-C25C00 ISERA111-C25D00 ISERA112-C29F30 ISERA111-C23Y00 ISERA111-C25X00 ISERA112-C23A30 ISERA112-C29B30 ISERA111-C23Q00 ISERA111-C29Z00 ISERA111-C25P00 ISERA111-C23U00 ISERA111-C29V00 ISERA111-C25T00
ISAspher HPLC Columns, C18 BDS
Colonnes pour chromatographie
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
C18-BDS columns with high C content, minimised silanol activity and additional shape selectivity (Polymer C18 bonding).
Certifications: Individual test certificate is enclosed with each column.
Information de livraison: Columns are delivered in solvent which is suitable for reversed phase chromatography.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité