ISAspher HPLC Columns, C8
Fournisseur: Isera
ISAspher
ISERA112-D25B00EA 251.5 EUR InStock
ISERA112-D25B00 ISERA113-D29L00 ISERA111-D29H00 ISERA112-D23C00 ISERA113-D29H00 ISERA113-D25J00 ISERA112-D25F00 ISERA111-D29L00 ISERA112-D23G00 ISERA111-D25J00 ISERA111-D29P00 ISERA111-D25N00 ISERA111-D29T00 ISERA111-D29X00 ISERA111-D25R00 ISERA111-D25V00 ISERA111-D25Z00 ISERA112-D29A30 ISERA112-D29E30 ISERA112-D25C30 ISERA112-D25G30 ISERA111-D23W00 ISERA111-D23S00 ISERA111-D23K00 ISERA111-D23G00 ISERA113-D29D00 ISERA111-D25F00 ISERA111-D23C00 ISERA113-D25B00 ISERA111-D29D00 ISERA111-D25B00 ISERA112-D25A00 ISERA112-D29C00 ISERA112-D25C00 ISERA113-D29K00 ISERA111-D29E00 ISERA112-D25G00 ISERA111-D29M00 ISERA111-D25K00 ISERA111-D29Q00 ISERA111-D29U00 ISERA111-D25S00 ISERA111-D29Y00 ISERA111-D25W00 ISERA112-D29B30 ISERA112-D23A30 ISERA112-D29F30 ISERA112-D23E30 ISERA111-D23X00 ISERA111-D23T00 ISERA111-D23P00 ISERA111-D23L00 ISERA111-D23H00 ISERA111-D25G00 ISERA111-D23D00 ISERA113-D25A00 ISERA111-D25C00 ISERA111-D29A00 ISERA111-D29F00 ISERA112-D23A00 ISERA112-D29F00 ISERA113-D25D00 ISERA111-D29J00 ISERA112-D23E00 ISERA113-D29J00 ISERA113-D25H00 ISERA111-D29N00 ISERA111-D25L00 ISERA113-D25L00 ISERA111-D29R00 ISERA111-D25P00 ISERA111-D25T00 ISERA111-D29V00 ISERA111-D25X00 ISERA111-D29Z00 ISERA112-D29C30 ISERA112-D29G30 ISERA112-D25A30 ISERA112-D23B30 ISERA112-D25E30 ISERA112-D23F30 ISERA111-D23Y00 ISERA111-D23U00 ISERA111-D23Q00 ISERA111-D23M00 ISERA111-D25H00 ISERA111-D23E00 ISERA112-D29A00 ISERA111-D29B00 ISERA111-D23A00 ISERA111-D25D00 ISERA112-D29E00 ISERA113-D29B00 ISERA112-D23B00 ISERA112-D25E00 ISERA112-D29G00 ISERA111-D29G00 ISERA112-D23F00 ISERA111-D29K00 ISERA113-D25K00 ISERA111-D25M00 ISERA111-D29S00 ISERA111-D29W00 ISERA111-D25Q00 ISERA111-D25U00 ISERA111-D25Y00 ISERA112-D25B30 ISERA112-D25F30 ISERA112-D23C30 ISERA112-D23G30 ISERA111-D23Z00 ISERA111-D23V00 ISERA111-D23R00 ISERA111-D23N00 ISERA111-D23J00 ISERA111-D23F00 ISERA111-D25E00 ISERA111-D23B00 ISERA112-D29B00 ISERA113-D29A00 ISERA111-D25A00 ISERA111-D29C00
ISAspher HPLC Columns, C8
Colonnes pour chromatographie
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
Certifications: Individual test certificate is enclosed with each column.
Information de livraison: Columns are delivered in solvent which is suitable for reversed phase chromatography.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité