ISAspher Reversed Phase HPLC Columns, Cyano
Fournisseur: Isera
ISAspher
ISERA111-H23V00EA 253 EUR InStock
ISERA111-H23V00 ISERA111-H25U00 ISERA111-H23Z00 ISERA111-H25Y00 ISERA112-H25F30 ISERA112-H25B30 ISERA112-H23C30 ISERA111-H23F00 ISERA112-H23F00 ISERA111-H25E00 ISERA111-H23J00 ISERA111-H29K00 ISERA111-H23N00 ISERA111-H25M00 ISERA111-H29W00 ISERA111-H23R00 ISERA111-H25Q00 ISERA111-H29S00 ISERA112-H29F00 ISERA111-H29G00 ISERA111-H23B00 ISERA112-H23A00 ISERA112-H29B00 ISERA111-H25A00 ISERA111-H29C00 ISERA112-H23G30 ISERA112-H29C30 ISERA111-H25X00 ISERA111-H23U00 ISERA111-H23Y00 ISERA111-H29Z00 ISERA112-H23B30 ISERA112-H25E30 ISERA112-H29G30 ISERA112-H25A30 ISERA112-H23F30 ISERA111-H29N00 ISERA111-H25H00 ISERA111-H23E00 ISERA112-H23E00 ISERA111-H29J00 ISERA111-H25L00 ISERA111-H25P00 ISERA111-H23M00 ISERA111-H29V00 ISERA111-H23Q00 ISERA111-H29R00 ISERA111-H25T00 ISERA112-H29A00 ISERA111-H29F00 ISERA112-H25C00 ISERA111-H23A00 ISERA111-H25D00 ISERA111-H29B00 ISERA112-H29E00 ISERA111-H25W00 ISERA112-H29B30 ISERA111-H23T00 ISERA111-H29Y00 ISERA111-H23X00 ISERA112-H23A30 ISERA112-H23E30 ISERA112-H29F30 ISERA111-H25G00 ISERA111-H29M00 ISERA111-H23D00 ISERA111-H25K00 ISERA112-H25G00 ISERA111-H23H00 ISERA111-H29U00 ISERA111-H23L00 ISERA111-H29Q00 ISERA111-H25S00 ISERA111-H23P00 ISERA112-H25B00 ISERA111-H29E00 ISERA112-H23C00 ISERA111-H29A00 ISERA111-H25C00 ISERA112-H29A30 ISERA111-H25V00 ISERA111-H23S00 ISERA111-H25Z00 ISERA111-H23W00 ISERA112-H25G30 ISERA112-H25C30 ISERA112-H29E30 ISERA112-H23G00 ISERA111-H29P00 ISERA111-H25F00 ISERA111-H23C00 ISERA112-H25F00 ISERA111-H29L00 ISERA111-H25J00 ISERA111-H23G00 ISERA111-H25N00 ISERA111-H29X00 ISERA111-H23K00 ISERA111-H25R00 ISERA111-H29T00 ISERA111-H29H00 ISERA112-H29G00 ISERA112-H25E00 ISERA112-H23B00 ISERA111-H25B00 ISERA111-H29D00 ISERA112-H29C00 ISERA112-H25A00
ISAspher Reversed Phase HPLC Columns, Cyano
Colonnes pour chromatographie
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
Certifications: Individual test certificate is enclosed with each column.
Information de livraison: Columns are delivered in solvent which is suitable for reversed phase / HILIC chromatography.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité