Manual Vial Crimping and Decapping Tongs
Fournisseur: ADELPHI MANUFACTURING
6030613 6030614 6030620 6030622 6030625 6030713 6030722 6030721
ADLM6030613EA 365.4 EUR InStock
ADLM6030613 ADLM6030614 ADLM6030620 ADLM6030622 ADLM6030625 ADLM6030713 ADLM6030722 ADLM6030721
Manual Vial Crimping and Decapping Tongs
Pinces à sertir et pinces à dessertir
A range of high-quality tongs and pliers for the crimping and decapping of pharmaceutical vials.
  • Options for 'plain', 'flip top/flip off', and 'flip tear up' seals
  • Crimping and decapping of 13 and 20 mm seals
  • Fast and reliable, for a professional quality finish
  • Easy, intuitive operation
All devices feature hardened, durable steel jaws to ensure reproducibility of seal crimp quality. They also feature an adjustable stop to ensure that equal pressure is applied to each crimp. Decapping pliers use a press and wrench action for removing seals from vials. The handles are plastic-coated for operator comfort.
Adelphi manufacturing also offers the semi-automatic powered vial crimper, to reduce operator fatigue.
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité