Syringe Stopper Press
Fournisseur: Adelphi Manufacturing
ADLMSP048-3001EA 5285.7 EUR InStock
ADLMSP048 ADLMSP048-3001
Syringe Stopper Press
Presses
Easy and efficient insertion of stoppers into pre-filled syringes of 6,35 mm bore or greater.
  • Easy and efficient insertion of stoppers into pre-filled syringes, ensuring that all air is expelled
  • Hygienic 316 L pharmaceutical grade stainless steel contact parts, to eliminate the risk of contamination
  • Fully adjustable design for operator comfort and safety
The unit comprises a syringe holding jig, lever operated stopper press, and a divide for aiding the removal of air from the syringe.
Informations de commande: Adelphi also offers the Accuramatic peristaltic pump (purchased separately), for filling nested syringes.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité