Steelite™ S2 Safety Shoes, Lace-Up, Low
Fournisseur: Portwest
Steelite™
PRTWFW80WHR49EA 45 EUR InStock
PRTWFW80WHR49
Steelite™ S2 Safety Shoes, Lace-Up, Low
Chaussures et bottes
Classic laced safety shoe with padded collar for all day comfort. Minimal design makes this shoe practical, safe, and durable.
  • Padded collar for added comfort
  • Wipe clean upper
  • Available in unisex sizing
  • Protective steel toecap
  • Anti-static footwear
  • Energy absorbing seat region
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel- and oil-resistant outsole
  • Mono density sole unit
The Steelite™ laced safety shoe features a self-cleaning outsole to prevent cross-contamination along with a water-resistant upper to prevent water penetration.
Certifications: EN ISO 20345: 2011 S2. CE certified.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité