Disposable Particle Filtering Respirators, FFP3, P371
Fournisseur: Portwest
PRTWP371WHREA 91.8 EUR InStock
PRTWP371WHR
Disposable Particle Filtering Respirators, FFP3, P371
Respirateurs Masques à usage unique Demi-masques fitrants contre les particules
Ergonomically designed to fit most face shapes, this respirator has a soft outer net cover to protect and retain its shape.
  • Ergonet supports the mask structure and keeps the filter media clean
  • Internal foam gasket ensures a tight face seal
  • Soft nose bridge for a secure and comfortable fit
  • Wide adjustable head straps for ultimate wearer comfort
  • Non-magnetic - nickel and ferrous free
  • Material: PP, EVA foam
High performance exhalation valve to reduce heat and provide comfort when used in hot and humid conditions. Dolomite optional clogging test (D) to provide more comfortable breathing and longer lasting filtration performance.
Its high performing exhalation valve works efficiently to reduce heat and moisture inside the mask and provides comfort when used in hot and humid conditions. Other features include an innovative moulded nose bridge for a perfect fit, all-round foam gasket and wide adjustable elastic head straps.
Certifications: EN 149:2001+A1:2009 FFP3 (NR D), CE-CAT III, UKCA marked.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité