Mitra® Microsamplers, Clamshell Format
Fournisseur: SGE a Trajan company
10001 20004
SGEA10001EA 0 EUR InStock
SGEA10001 SGEA20004
Mitra® Microsamplers, Clamshell Format
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
The Mitra® is a small, portable device with an absorbent VAMS® tip that is designed to collect a tiny, precise volume of blood. The Mitra device is used for capillary sampling from a fingertip or other site on the body. This approach is less invasive than venipuncture, doesn't require expensive cold shipping, and requires only a small blood volume for dried-matrix analysis.
  • Number of samples collected: Up to four
  • Substrate: Polymer
  • Shelf life: 1,5 years
  • Volumetric precision (%RSD): <5%
Mitra® devices are intended as a single-use, specimen collector and for the storage and transport of blood.
For transport of samples from end-user to the lab, the desiccant contained in the sealed specimen bag will keep the samples dry during transit. For long-term storage in a deep freezer of sampled Mitra devices (clamshells or
cartridges).
The samples can be placed inside a large storage bag containing desiccant, which keeps the samples dry
during cold storage in the lab. The zip closure prevents moisture from entering the storage bag. A label on the outside of both the individual specimen bag and the larger storage bag enables samples to be easily marked and identified.
Certifications: Mitra® devices are Class 1 IVD in Australia, Brazil & China, Class B in South Africa, and registered with health agencies in Canada and Ukraine.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité