FEATHER® Scalpel Blades
Fournisseur: PFM Medical
Feather®
FEAT201000370EA 109 EUR InStock
FEAT201000370
FEATHER® Scalpel Blades
Bistouris et pièces détachées Lames de bistouri
FEATHER® scalpel blades fig. 370 are specifically suitable for surgical applications in dental surgery.
  • Blade: Stainless steel
Precise cutting and performance: The stainless steel blades offer outstanding sharpness and superb durability due to double-honing precision grinding.
Particularly compatible: The blades are suitable for all standard surgical handles (Size 3 and 7) as well as with the FEATHER® Titan Roundshaft Handle.
User friendly: The blades are suitable for applications under the microscope and are ready for use immediately after removal from the sterile packaging.
Safe application for horizontal enlargement incisions of flaps. Suitable for incisions in regenerative surgery.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité