Conjugation kits, protein-protein conjugation kit
Fournisseur: Solulink
SOLUS-9010-1EA 0 EUR InStock
SOLUS-9010-1
Conjugation kits, protein-protein conjugation kit
Tests Essais protéiniques
The Protein-Protein Conjugation kit is designed to easily and efficiently conjugate two proteins together. This kit is flexible so that researchers with little or no conjugation experience can make their own custom protein-protein conjugates to suit their needs. It includes all of the necessary components - including S-HyNic and S-4FB - and protocols for easy and specific crosslinking of any protein with any other protein of a different size greater than 7 kDa.
  • Catalysed conjugation – Faster kinetics for greater efficiency and yields
  • Quantifiable – Using a UV signature wavelength and simple UV scan
  • Stability – 10 times more stable than any other conjugation linker
  • Specificity – Two-linker method avoids homoconjugate formation
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité