AATB11067EA
280
EUR
InStock
AATB11067
XFD350 tyramide
XFD350 tyramide
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
- Bright signal
- Excellent signal amplification for low expression targets