AATB13001EA
739
EUR
InStock
AATB13001
BOC-Leu-Gly-Arg-Aminoluciferin
BOC-Leu-Gly-Arg-Aminoluciferin
This aminoluciferin-based probe is a luminogenic substrate for complement component C3/C5 convertases, coagulation factor Xa, and soybean trypsin-like enzyme.
- High sensitivity
- Excellent quality luminogenic substrate
- Adaptable to multiple types of luciferase assays