AATB13467EA
146
EUR
InStock
AATB13467
(Ac-PAL)2R110
(Ac-PAL)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
- Protease substrate
- Bright signal
- Multiple applications