AATB13481EA
442
EUR
InStock
AATB13481
Bis-Arg-rhodamine 110
Bis-Arg-rhodamine 110
This Bis Arg-rhodamine 110 is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of human alpha-thrombin.
- Alpha-thrombin substrate
- Bright signal
- Multiple applications