AATB2076EA
295
EUR
InStock
AATB2076
Tide Quencher™ 5WS amine, Tide Quencher™ sonde fluorescente
Tide Quencher™ 5WS amine
TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling