AATB2236EA
295
EUR
InStock
AATB2236
Tide Fluor™ 1 azide, Tide Fluor™ sonde fluorescente
Tide Fluor™ 1 azide
TF1 is designed to be a superior fluorophore alternative to EDANS.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity