AATB2248EA
295
EUR
InStock
AATB2248
Tide Fluor™ 2, succinimidyl ester, Tide Fluor™ sonde fluorescente
Tide Fluor™ 2, succinimidyl ester
Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity