AATB4530EA
146
EUR
InStock
AATB4530
DBCO-PEG4-NHS Ester for biomolecular labeling
DBCO-PEG4-NHS Ester
Cu-free click reaction, i.e., the strain-promoted alkyne-azide cycloaddition (SPAAC) is a bioorthogonal reaction utilizing a pair of reagents, cyclooctynes and azides that exclusively and efficiently react with each other while remain inert to naturally occurring functional groups such as hydroxy, amino and thiol groups.
- Improved crosslinking efficiency
- High water solubility