Mag-Fluo-4, Tetrapotassium Salt
Fournisseur: Biotium
50047.EA 0 EUR InStock
50047.
Mag-Fluo-4, Tetrapotassium Salt
Mag-Fluo-4, Tetrapotassium Salt
Mag-Fluo-4 is a fluorescent ion indicator and analog of Fluo-4. The indicator has an affinity for magnesium (Kd = 4,7 mM) and low-affinity for calcium (Kd = 22 µm) with absorbance/emission at 493/517 nm (high Mg2+). The cell membrane-impermeant tetrapotassium salt form can be introduced into cells by microinjection.
  • λEx/λEm (high Mg2+): 493/517 nm
  • Orange solid soluble in water
  • Store 4 °C and protect from light
BAPTA-based ion indicators like Mag-Fluo-4 have been shown to be fixable in situ by EDC/EDAC (cat# 59002). The fixation of indicator dyes is useful for downstream immunofluorescence and IHC studies (Cell Calcium 1997, 21(3), 175).
As the indicator does not covalently bind to cellular components, it may be actively effluxed from the cell by organic anion transporters. The rate of efflux increases with temperature, and may vary between cell types, resulting in variable retention times of a few minutes to hours. Experiments using indicators in cells usually are performed within one or two hours of loading, but it may be possible to re-load cells with indicator if needed. The organic anion transporter inhibitor Probenecid can be used to slow the rate of indicator efflux from cells.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité