Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Fournisseur: AAT Bioquest
Bucculite™
AATB22326EA 683 EUR InStock
AATB22326
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Tests Tests cellulaires
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • FITC filter set compatible
  • Bright fluorescence
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit uses XdU which is incorporated into cellular DNA during DNA synthesis. After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is imaged with a FITC filter set. Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay..
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité