Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Fournisseur: AAT Bioquest
Bucculite™
AATB22323EA 683 EUR InStock
AATB22323
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Tests Tests cellulaires
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • Blue laser compatible
  • Highly reliable
  • Optimised for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is visualised in FITC Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité