Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Fournisseur: AAT Bioquest
Bucculite™
AATB22321EA 683 EUR InStock
AATB22321
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Tests Tests cellulaires
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • Violet laser compatible
  • Highly reliable
  • Optimised for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with mFluor™ Violet 450 MTA. The resulted mFluor™ Violet 450-labeled DNA formed in cells is visualised in Pacific Blue Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay..
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité