AATB11066EA
146
EUR
InStock
AATB11066
Cy5 tyramide
Cy5 tyramide
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
- Bright signal
- Excellent signal amplification for low expression targets